Assay Design
Gene Transcripts
- Gene Symbol
- TRIM55
- Entrez Gene ID
-
84675
- Gene Name
- tripartite motif containing 55
- Gene Aliases
-
MURF-2, RNF29, muRF2
- Chromosome Location
-
Chr.8: 66112667 - 66175485 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_184087.1 |
NP_908975.1 |
4 - 5 |
822 |
75 |
GenBank mRNA |
AJ277493.1 |
- |
4 - 5 |
822 |
75 |
View Full Product Details