Assay Design



Gene Transcripts

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
-
Chromosome Location
Chr.4: 119296419 - 119304445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170330.1 NP_001163801.1 1 - 2 201 81
GenBank mRNA AY605045.1 - 1 - 2 201 81

View Full Product Details