Assay Design



Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr.X: 2903970 - 2929375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001669.3 NP_001660.2 4 - 5 539 59
NM_009589.3 NP_033667.2 4 - 5 539 59
XM_005274514.2 XP_005274571.1 4 - 5 509 59
XM_005274515.2 XP_005274572.1 4 - 5 509 59
GenBank mRNA AF160499.1 - 4 - 5 439 59
AK314479.1 - 4 - 5 503 59
BC020229.1 - - 467 59
X83572.1 - 4 - 5 515 59

View Full Product Details