Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014299.2 | NP_055114.1 | 5 - 6 | 1067 | 77 |
NM_058243.2 | NP_490597.1 | 5 - 6 | 1067 | 77 | |
XM_011527854.1 | XP_011526156.1 | 5 - 6 | 1132 | 77 | |
XM_011527855.1 | XP_011526157.1 | 5 - 6 | 1133 | 77 | |
XM_011527856.2 | XP_011526158.1 | 5 - 6 | 1132 | 77 | |
GenBank mRNA | AF386649.1 | - | 5 - 6 | 1067 | 77 |
AK307664.1 | - | 5 - 6 | 957 | 77 | |
BC000156.1 | - | 5 - 6 | 913 | 77 | |
BC030158.2 | - | 5 - 6 | 934 | 77 | |
BC035266.1 | - | 5 - 6 | 1138 | 77 | |
BC038988.1 | - | 5 - 6 | 1108 | 77 | |
BC047888.1 | - | 5 - 6 | 1121 | 77 | |
BC067129.1 | - | 5 - 6 | 1089 | 77 | |
BC091649.1 | - | 5 - 6 | 1105 | 77 | |
Y12059.1 | - | 5 - 6 | 1067 | 77 |