Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317177.1 | NP_001304106.1 | 8 - 9 | 895 | 97 |
NM_017812.3 | NP_060282.1 | 7 - 8 | 880 | 97 | |
NR_133671.1 | 10 - 11 | 1261 | 97 | ||
GenBank mRNA | AK000427.1 | - | 7 - 8 | 796 | 97 |
AK129533.1 | - | 7 - 8 | 761 | 97 | |
AK310236.1 | - | 10 - 11 | 1173 | 97 | |
BC011596.2 | - | 7 - 8 | 802 | 97 | |
BC014839.1 | - | 7 - 8 | 804 | 97 |