Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053053.3 | NP_444281.1 | 3 - 4 | 410 | 65 |
GenBank mRNA | AK290964.1 | - | 3 - 4 | 227 | 65 |
AK291922.1 | - | 3 - 4 | 350 | 65 | |
AK309250.1 | - | 3 - 4 | 278 | 65 | |
BC015401.2 | - | 3 - 4 | 235 | 65 | |
CN399642.1 | - | 3 - 4 | 412 | 65 |