Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267798.1 | NP_001254727.1 | 2 - 3 | 280 | 57 |
NM_002838.4 | NP_002829.3 | 2 - 3 | 280 | 57 | |
NM_080921.3 | NP_563578.2 | 2 - 3 | 280 | 57 | |
NR_052021.1 | 2 - 3 | 280 | 57 | ||
XM_006711472.3 | XP_006711535.1 | 2 - 3 | 241 | 57 | |
XM_006711473.3 | XP_006711536.1 | 2 - 3 | 223 | 57 | |
XM_006711474.3 | XP_006711537.1 | 2 - 3 | 223 | 57 | |
GenBank mRNA | AK130573.1 | - | 2 - 3 | 153 | 57 |
AK292131.1 | - | 2 - 3 | 153 | 57 | |
AK299986.1 | - | 2 - 3 | 153 | 57 | |
AK303539.1 | - | 2 - 3 | 153 | 57 | |
AK307635.1 | - | 2 - 3 | 158 | 57 | |
AK308217.1 | - | 2 - 3 | 229 | 57 | |
AK310898.1 | - | 2 - 3 | 176 | 57 | |
AY567999.1 | - | 1 - 2 | 81 | 57 | |
BC014239.2 | - | 2 - 3 | 144 | 57 | |
BC017863.1 | - | 2 - 3 | 152 | 57 | |
BC121086.1 | - | 1 - 2 | 90 | 57 | |
BC121087.1 | - | 1 - 2 | 90 | 57 | |
BC127656.1 | - | 1 - 2 | 90 | 57 | |
BC127657.1 | - | 1 - 2 | 90 | 57 | |
BC148257.1 | - | 1 - 2 | 90 | 57 | |
DA670254.1 | - | 2 - 3 | 183 | 57 | |
DA819314.1 | - | 2 - 3 | 280 | 57 | |
DA948670.1 | - | 2 - 3 | 243 | 57 | |
Y00062.1 | - | 3 - 4 | 207 | 57 | |
Y00638.1 | - | 2 - 3 | 147 | 57 |