Assay Design



Gene Transcripts

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
Chromosome Location
Chr.X: 118495664 - 118686162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144658.3 NP_653259.3 14 - 15 1663 94
XM_005262368.3 XP_005262425.1 14 - 15 1908 94
XM_005262369.4 XP_005262426.1 14 - 15 1602 94
XM_011531276.2 XP_011529578.1 14 - 15 1543 94
GenBank mRNA AK124735.1 - 8 - 9 1048 94
AY692226.1 - 14 - 15 1663 94
BF513250.1 - 3 - 4 94

View Full Product Details