Assay Design



Gene Transcripts

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
Chromosome Location
Chr.X: 118495664 - 118686162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144658.3 NP_653259.3 30 - 31 3352 103
GenBank mRNA AK125641.1 - 9 - 10 864 103
AY692226.1 - 30 - 31 3352 103

View Full Product Details