Assay Design
Gene Transcripts
- Gene Symbol
- AGAP5
- Entrez Gene ID
-
729092
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
- Gene Aliases
-
CTGLF2
- Chromosome Location
-
Chr.10: 73674275 - 73697796 on Build GRCh38
- Gene Symbol
- AGAP12P
- Entrez Gene ID
-
414224
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 12, pseudogene
- Gene Aliases
-
CTGLF12P
- Chromosome Location
-
Chr.10: 48010132 - 48031640 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029396.2 |
|
2 - 3 |
340 |
74 |
- Gene Symbol
- AGAP4
- Entrez Gene ID
-
119016
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
- Gene Aliases
-
AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
- Chromosome Location
-
Chr.10: 45825592 - 45858529 on Build GRCh38
- Gene Symbol
- AGAP9
- Entrez Gene ID
-
642517
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
- Gene Aliases
-
AGAP-9, CTGLF6, bA301J7.2
- Chromosome Location
-
Chr.10: 47501854 - 47523638 on Build GRCh38
- Gene Symbol
- LOC107984026
- Entrez Gene ID
-
107984026
- Gene Name
- arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 46335552 - 46838268 on Build GRCh38
View Full Product Details