Assay Design
Gene Transcripts
- Gene Symbol
- SPATA31D4
- Entrez Gene ID
-
389761
- Gene Name
- SPATA31 subfamily D member 4
- Gene Aliases
-
FAM75D4
- Chromosome Location
-
Chr.9: 81928428 - 81934998 on Build GRCh38
- Gene Symbol
- SPATA31D3
- Entrez Gene ID
-
389762
- Gene Name
- SPATA31 subfamily D member 3
- Gene Aliases
-
FAM75D3
- Chromosome Location
-
Chr.9: 81943500 - 81950094 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207416.2 |
NP_997299.2 |
2 - 3 |
315 |
85 |
View Full Product Details