Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317.5 | NP_001308.1 | 2 - 3 | 319 | 85 |
GenBank mRNA | AK291587.1 | - | 2 - 3 | 238 | 85 |
AU134786.2 | - | 2 - 3 | 319 | 85 | |
BC002717.2 | - | 2 - 3 | 212 | 85 | |
BC005921.1 | - | 2 - 3 | 231 | 85 | |
BC020756.1 | - | 2 - 3 | 287 | 85 | |
BC057768.1 | - | 2 - 3 | 231 | 85 | |
BC062775.1 | - | 2 - 3 | 235 | 85 | |
BG436049.1 | - | 2 - 3 | 232 | 85 | |
BT006926.1 | - | 1 - 2 | 169 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020991.3 | NP_066271.1 | 2 - 3 | 284 | 85 |
NM_022644.3 | NP_072170.1 | 2 - 3 | 284 | 85 | |
GenBank mRNA | BC022044.1 | - | 2 - 3 | 235 | 85 |
BC035965.1 | - | 2 - 3 | 231 | 85 | |
BC119748.2 | - | 2 - 3 | 228 | 85 | |
V00573.1 | - | 2 - 3 | 193 | 85 |