Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317.5 | NP_001308.1 | 3 - 4 | 442 | 59 |
GenBank mRNA | AK291587.1 | - | 3 - 4 | 361 | 59 |
AU134786.2 | - | 3 - 4 | 442 | 59 | |
BC002717.2 | - | 3 - 4 | 335 | 59 | |
BC005921.1 | - | 3 - 4 | 354 | 59 | |
BC020756.1 | - | 3 - 4 | 410 | 59 | |
BC057768.1 | - | 3 - 4 | 354 | 59 | |
BC062775.1 | - | 3 - 4 | 358 | 59 | |
BG436049.1 | - | 3 - 4 | 355 | 59 | |
BT006926.1 | - | 2 - 3 | 292 | 59 | |
J00118.1 | - | 3 - 4 | 321 | 59 | |
V00593.1 | - | 2 - 3 | 144 | 59 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020991.3 | NP_066271.1 | 3 - 4 | 407 | 59 |
NM_022644.3 | NP_072170.1 | 3 - 4 | 407 | 59 | |
GenBank mRNA | BC022044.1 | - | 3 - 4 | 358 | 59 |
BC035965.1 | - | 3 - 4 | 354 | 59 | |
BC119748.2 | - | 3 - 4 | 351 | 59 | |
V00573.1 | - | 3 - 4 | 316 | 59 |