Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317.5 | NP_001308.1 | 4 - 5 | 608 | 101 |
GenBank mRNA | AK291587.1 | - | 4 - 5 | 527 | 101 |
BC002717.2 | - | 4 - 5 | 501 | 101 | |
BC005921.1 | - | 4 - 5 | 520 | 101 | |
BC020756.1 | - | 4 - 5 | 576 | 101 | |
BC057768.1 | - | 4 - 5 | 520 | 101 | |
BC062775.1 | - | 4 - 5 | 524 | 101 | |
BG436049.1 | - | 4 - 5 | 521 | 101 | |
BQ004002.1 | - | 1 - 2 | 101 | ||
BT006926.1 | - | 3 - 4 | 458 | 101 | |
J00118.1 | - | 4 - 5 | 487 | 101 | |
V00593.1 | - | 3 - 4 | 310 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020991.3 | NP_066271.1 | 4 - 5 | 573 | 101 |
GenBank mRNA | BC022044.1 | - | 4 - 5 | 524 | 101 |
BC035965.1 | - | 4 - 5 | 520 | 101 | |
BC119748.2 | - | 4 - 5 | 517 | 101 | |
V00573.1 | - | 4 - 5 | 482 | 101 |