Assay Design
Gene Transcripts
- Gene Symbol
- NBPF11
- Entrez Gene ID
-
200030
- Gene Name
- neuroblastoma breakpoint family member 11
- Gene Aliases
-
NBPF24
- Chromosome Location
-
Chr.1: 148102046 - 148152190 on Build GRCh38
- Gene Symbol
- LOC102724250
- Entrez Gene ID
-
102724250
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1|NT_187366: 4611 - 41174 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_011546188.2 |
XP_011544490.1 |
15 - 16 |
10767 |
104 |
- Gene Symbol
- NBPF1
- Entrez Gene ID
-
55672
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
-
AB13, AB14, AB23, AD2, NBG, NBPF
- Chromosome Location
-
Chr.1: 16562427 - 16613605 on Build GRCh38
- Gene Symbol
- NBPF10
- Entrez Gene ID
-
100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
-
AB6, AG1, NBPF9
- Chromosome Location
-
Chr.1: 146066318 - 146144804 on Build GRCh38
- Gene Symbol
- NBPF14
- Entrez Gene ID
-
25832
- Gene Name
- neuroblastoma breakpoint family member 14
- Gene Aliases
-
DJ328E19.C1.1, NBPF
- Chromosome Location
-
Chr.1: 148531385 - 148607636 on Build GRCh38
- Gene Symbol
- NBPF15
- Entrez Gene ID
-
284565
- Gene Name
- neuroblastoma breakpoint family member 15
- Gene Aliases
-
AB14, AG3, NBPF16
- Chromosome Location
-
Chr.1: 144421386 - 144459464 on Build GRCh38
- Gene Symbol
- NBPF19
- Entrez Gene ID
-
101060226
- Gene Name
- neuroblastoma breakpoint family member 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 149472955 - 149556256 on Build GRCh38
- Gene Symbol
- NBPF8
- Entrez Gene ID
-
728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
-
NBPF8P
- Chromosome Location
-
Chr.1: 120436351 - 120467844 on Build GRCh38
- Gene Symbol
- NBPF9
- Entrez Gene ID
-
400818
- Gene Name
- neuroblastoma breakpoint family member 9
- Gene Aliases
-
AE01
- Chromosome Location
-
Chr.1: 149054033 - 149090861 on Build GRCh38
View Full Product Details