Assay Design
Gene Transcripts
- Gene Symbol
- NBPF8
- Entrez Gene ID
-
728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
-
NBPF8P
- Chromosome Location
-
Chr.1: 120436351 - 120467844 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY894569.1 |
- |
2 - 3 |
237 |
102 |
- Gene Symbol
- NBPF11
- Entrez Gene ID
-
200030
- Gene Name
- neuroblastoma breakpoint family member 11
- Gene Aliases
-
NBPF24
- Chromosome Location
-
Chr.1: 148102046 - 148152190 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110864.1 |
|
5 - 6 |
1203 |
102 |
GenBank mRNA |
AF419617.1 |
- |
5 - 6 |
706 |
102 |
BX648497.1 |
- |
5 - 6 |
1002 |
102 |
View Full Product Details