Assay Design
Gene Transcripts
- Gene Symbol
- MT2A
- Entrez Gene ID
-
4502
- Gene Name
- metallothionein 2A
- Gene Aliases
-
MT2
- Chromosome Location
-
Chr.16: 56608566 - 56609497 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_005953.3 |
NP_005944.1 |
1 - 1 |
54 |
84 |
GenBank mRNA |
BM746626.1 |
- |
1 - 1 |
54 |
84 |
View Full Product Details