Assay Design



Gene Transcripts

Gene Symbol
HLA-G
Entrez Gene ID
3135
Gene Name
major histocompatibility complex, class I, G
Gene Aliases
MHC-G
Chromosome Location
Chr.6: 29826967 - 29831130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002127.5 NP_002118.1 1 - 1 50 85
GenBank mRNA AK302054.1 - 1 - 1 50 85
DC397107.1 - 1 - 1 50 85

View Full Product Details