Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010.2 | NP_001001.2 | 4 - 4 | 454 | 106 |
GenBank mRNA | AB062123.1 | - | 4 - 4 | 440 | 106 |
AK093634.1 | - | - | 1655 | 106 | |
AK291517.1 | - | 4 - 4 | 453 | 106 | |
AK311861.1 | - | 4 - 4 | 451 | 106 | |
BC000524.2 | - | 3 - 3 | 416 | 106 | |
BC009427.2 | - | 3 - 3 | 407 | 106 | |
BC013296.2 | - | 4 - 4 | 437 | 106 | |
BC027620.1 | - | 4 - 4 | 457 | 106 | |
BC035447.1 | - | 2 - 2 | 278 | 106 | |
BC071907.1 | - | 4 - 4 | 442 | 106 | |
BC071908.1 | - | 4 - 4 | 442 | 106 | |
BC094826.1 | - | 4 - 4 | 438 | 106 | |
M20020.1 | - | 4 - 4 | 439 | 106 |