Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097622.1 | NP_001091091.1 | 1 - 1 | 48 | 112 |
GenBank mRNA | BC113704.1 | - | 1 - 1 | 48 | 112 |
BC113706.1 | - | 1 - 1 | 48 | 112 | |
BC147028.1 | - | 1 - 1 | 50 | 112 | |
BC147029.1 | - | 1 - 1 | 50 | 112 |