Assay Design
Gene Transcripts
- Gene Symbol
- MIRLET7C
- Entrez Gene ID
-
406885
- Gene Name
- microRNA let-7c
- Gene Aliases
-
LET7C, MIRNLET7C, hsa-let-7c, let-7c
- Chromosome Location
-
Chr.21: 16539828 - 16539911 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029480.1 |
|
1 - 1 |
44 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 99a |
MIR99A |
MIRN99A, mir-99a |
-659.0
|
Sense |
Chr. - 16539089 - 16539169 on Build GRCh38 |
Downstream |
microRNA 125b-2 |
MIR125B2 |
MIRN125B2, mir-125b-2 |
50.3kb
|
Sense |
Chr. - 16590237 - 16590325 on Build GRCh38 |
View Full Product Details