Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_029513.1 | 1 - 1 | 57 | 94 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase | HUWE1 | ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1 | Sense | Chr. - 53532096 - 53686729 on Build GRCh38 | |
Downstream | microRNA let-7f-2 | MIRLET7F2 | LET7F2, MIRNLET7F2, let-7f-2 | 851.0 | Sense | Chr. - 53557192 - 53557274 on Build GRCh38 |