Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_036101.1 | 1 - 1 | 41 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | Fer3-like bHLH transcription factor | FERD3L | N-TWIST, NATO3, NTWIST, bHLHa31 | -559.9kb | Sense | Chr. - 19144782 - 19145421 on Build GRCh38 |
Downstream | transmembrane protein 196 | TMEM196 | 13.9kb | Sense | Chr. - 19719315 - 19773700 on Build GRCh38 |