Assay Design
Gene Transcripts
- Gene Symbol
- DPY19L1P1
- Entrez Gene ID
-
100129460
- Gene Name
- DPY19L1 pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 32580939 - 32719168 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_036680.1 |
|
14 - 15 |
1276 |
89 |
GenBank mRNA |
AK026768.1 |
- |
14 - 15 |
1279 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated |
LSM5 |
YER146W |
-90.7kb
|
Sense |
Chr. - 32485333 - 32495258 on Build GRCh38 |
Downstream |
zinc and ring finger 2 pseudogene 1 |
ZNRF2P1 |
|
136.9kb
|
Anti-sense |
Chr. - 32727950 - 32729983 on Build GRCh38 |
View Full Product Details