Assay Design
Gene Transcripts
- Gene Symbol
- ATP1A1-AS1
- Entrez Gene ID
-
84852
- Gene Name
- ATP1A1 antisense RNA 1
- Gene Aliases
-
ATP1A1OS, C1orf203
- Chromosome Location
-
Chr.1: 116392865 - 116418622 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX335080.2 |
- |
1 - 2 |
99 |
|
Chr. 1 - 116398669 - 116418562 [-] on Build GRCh38 |
85 |
GenBankmRNA |
CA433193.1 |
- |
2 - 3 |
|
|
Chr. 1 - 116398999 - 116418536 [+] on Build GRCh38 |
85 |
GenBankmRNA |
BI827290.1 |
- |
1 - 2 |
112 |
|
Chr. 1 - 116399000 - 116418574 [-] on Build GRCh38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
mab-21 like 3 |
MAB21L3 |
C1orf161 |
-265.7kb
|
Anti-sense |
Chr. - 116072744 - 116135240 on Build GRCh38 |
Downstream |
uncharacterized LOC101929023 |
LOC101929023 |
|
5.1kb
|
Sense |
Chr. - 116423724 - 116478826 on Build GRCh38 |
View Full Product Details