Assay Design
Gene Transcripts
- Gene Symbol
- TMEM30C
- Entrez Gene ID
-
644444
- Gene Name
- transmembrane protein 30C
- Gene Aliases
-
CDC50C
- Chromosome Location
-
Chr.3: 100185824 - 100194186 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cms1 ribosomal small subunit homolog (yeast) |
CMSS1 |
C3orf26 |
-7.2kb
|
Sense |
Chr. - 99817834 - 100178632 on Build GRCh38 |
Downstream |
TBC1 domain family member 23 |
TBC1D23 |
NS4ATP1 |
70.6kb
|
Sense |
Chr. - 100260817 - 100325252 on Build GRCh38 |
View Full Product Details