Assay Design
Gene Transcripts
- Gene Symbol
- OR2M1P
- Entrez Gene ID
-
388762
- Gene Name
- olfactory receptor family 2 subfamily M member 1 pseudogene
- Gene Aliases
-
JCG10, OR2M1, OST037
- Chromosome Location
-
Chr.1: 248122136 - 248122780 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002141.1 |
|
1 - 1 |
550 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor family 2 subfamily L member 13 |
OR2L13 |
OR2L14 |
-21.2kb
|
Sense |
Chr. - 247937029 - 248100922 on Build GRCh38 |
Downstream |
olfactory receptor family 2 subfamily M member 5 |
OR2M5 |
OR2M5P |
22.4kb
|
Sense |
Chr. - 248145148 - 248146086 on Build GRCh38 |
View Full Product Details