Assay Design



Gene Transcripts

Gene Symbol
MCIDAS
Entrez Gene ID
345643
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Gene Aliases
IDAS, MCI, MCIN
Chromosome Location
Chr.5: 55219597 - 55227315 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190787.1 NP_001177716.1 1 - 2 294 82
GenBank mRNA FR854393.1 - 1 - 2 117 82

View Full Product Details