Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001818.3 | NP_001809.3 | 3 - 4 | 404 | 103 |
GenBank mRNA | AB031085.1 | - | 3 - 4 | 371 | 103 |
AK314988.1 | - | 3 - 4 | 445 | 103 | |
BC020744.1 | - | 3 - 4 | 404 | 103 | |
D26125.1 | - | 3 - 4 | 367 | 103 | |
M33375.1 | - | 3 - 4 | 369 | 103 | |
S68287.1 | - | 3 - 4 | 378 | 103 |