Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006418.4 | NP_006409.3 | 4 - 5 | 823 | 104 |
GenBank mRNA | AF097021.1 | - | 4 - 5 | 740 | 104 |
AK300947.1 | - | 4 - 5 | 703 | 104 | |
AY358567.1 | - | 4 - 5 | 740 | 104 | |
BC047740.1 | - | 3 - 4 | 454 | 104 | |
BC117329.1 | - | 4 - 5 | 805 | 104 |