Assay Design



Gene Transcripts

Gene Symbol
TCTEX1D2
Entrez Gene ID
255758
Gene Name
Tctex1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 196291219 - 196318294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152773.4 NP_689986.2 4 - 5 524 92
GenBank mRNA BC021177.2 - 4 - 5 504 92
BM548106.1 - 5 - 6 667 92
BM983853.1 - - 92

View Full Product Details