Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145873.1 | NP_001139345.1 | 9 - 9 | 1757 | 82 |
NM_001768.6 | NP_001759.3 | 6 - 6 | 1615 | 82 | |
NM_171827.3 | NP_741969.1 | 5 - 5 | 1504 | 82 | |
NR_027353.1 | 7 - 7 | 1201 | 82 | ||
GenBank mRNA | AK097942.1 | - | 7 - 7 | 800 | 82 |
AK124156.1 | - | 9 - 9 | 1757 | 82 | |
AK300089.1 | - | 7 - 7 | 1167 | 82 | |
AY039664.1 | - | 6 - 6 | 905 | 82 | |
BC025715.1 | - | 6 - 6 | 731 | 82 | |
M12824.1 | - | 6 - 6 | 813 | 82 | |
M12828.1 | - | 6 - 6 | 791 | 82 |