Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145873.1 | NP_001139345.1 | 9 - 9 | 2261 | 66 |
NM_001768.6 | NP_001759.3 | 6 - 6 | 2119 | 66 | |
NM_171827.3 | NP_741969.1 | 5 - 5 | 2008 | 66 | |
NR_027353.1 | 7 - 7 | 1705 | 66 | ||
GenBank mRNA | AK097942.1 | - | 7 - 7 | 1304 | 66 |
AK124156.1 | - | 9 - 9 | 2261 | 66 | |
BC025715.1 | - | 6 - 6 | 1235 | 66 | |
M12824.1 | - | 6 - 6 | 1317 | 66 |