Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297698.1 | NP_001284627.1 | 4 - 4 | 1653 | 99 |
NM_024865.3 | NP_079141.2 | 4 - 4 | 1701 | 99 | |
GenBank mRNA | AB093576.1 | - | 4 - 4 | 121 | |
AK022643.1 | - | 4 - 4 | 121 | ||
BC069807.1 | - | 1 - 1 | 1651 | 121 |