Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000142.4 | NP_000133.1 | 7 - 8 | 1184 | 89 |
XM_006713872.1 | XP_006713935.1 | 7 - 8 | 1197 | 89 | |
XM_006713873.1 | XP_006713936.1 | 7 - 8 | 1197 | 89 | |
XM_011513420.1 | XP_011511722.1 | 7 - 8 | 1197 | 89 | |
XM_011513422.1 | XP_011511724.1 | 7 - 8 | 1197 | 89 | |
GenBank mRNA | AB209441.1 | - | 7 - 8 | 1189 | 89 |
KJ534834.1 | - | 6 - 7 | 928 | 89 | |
M58051.1 | - | 6 - 7 | 967 | 89 | |
M64347.1 | - | 5 - 6 | 703 | 89 |