Assay Design
Gene Transcripts
- Gene Symbol
- ATP5O
- Entrez Gene ID
-
539
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
- Gene Aliases
-
ATPO, HMC08D05, OSCP
- Chromosome Location
-
Chr.21: 33903453 - 33915854 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001697.2 |
NP_001688.1 |
1 - 1 |
76 |
106 |
GenBank mRNA |
AV734214.1 |
- |
1 - 1 |
76 |
106 |
View Full Product Details