Assay Design
Gene Transcripts
- Gene Symbol
- MIR4526
- Entrez Gene ID
-
100616130
- Gene Name
- microRNA 4526
- Gene Aliases
-
mir-4526
- Chromosome Location
-
Chr.18: 13611114 - 13611200 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_039752.1 |
|
1 - 1 |
41 |
56 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 5190 |
MIR5190 |
|
-151.1kb
|
Sense |
Chr. - 13459947 - 13460026 on Build GRCh38 |
Downstream |
family with sequence similarity 210 member A |
FAM210A |
C18orf19, HsT2329 |
52.1kb
|
Anti-sense |
Chr. - 13663347 - 13726592 on Build GRCh38 |
View Full Product Details