Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161009.2 | NP_001154481.1 | 2 - 3 | 685 | 91 |
NM_001161010.2 | NP_001154482.1 | 3 - 4 | 790 | 91 | |
NM_001161011.2 | NP_001154483.1 | 2 - 3 | 685 | 91 | |
NM_001161012.2 | NP_001154484.1 | 2 - 3 | 685 | 91 | |
NM_001161013.2 | NP_001154485.1 | 2 - 3 | 685 | 91 | |
NM_031905.4 | NP_114111.2 | 3 - 4 | 790 | 91 | |
XM_005250628.2 | XP_005250685.1 | 3 - 4 | 944 | 91 | |
XM_011516601.2 | XP_011514903.1 | 2 - 3 | 863 | 91 | |
XM_011516602.2 | XP_011514904.1 | 2 - 3 | 224 | 91 | |
XM_017012684.1 | XP_016868173.1 | 4 - 5 | 971 | 91 | |
XM_017012685.1 | XP_016868174.1 | 4 - 5 | 971 | 91 | |
XM_017012686.1 | XP_016868175.1 | 4 - 5 | 971 | 91 | |
XM_017012687.1 | XP_016868176.1 | 2 - 3 | 203 | 91 | |
XM_017012688.1 | XP_016868177.1 | 4 - 5 | 971 | 91 | |
GenBank mRNA | AF275808.1 | - | 2 - 3 | 193 | 91 |
AK075500.1 | - | 3 - 4 | 531 | 91 | |
AK225082.1 | - | 2 - 3 | 418 | 91 | |
AK291818.1 | - | 2 - 3 | 413 | 91 | |
AK301565.1 | - | 2 - 3 | 418 | 91 | |
AY150851.1 | - | 2 - 3 | 685 | 91 | |
AY150852.1 | - | 3 - 4 | 790 | 91 | |
AY150853.1 | - | 2 - 3 | 685 | 91 | |
AY150854.1 | - | 3 - 4 | 790 | 91 | |
BC003586.1 | - | 2 - 3 | 405 | 91 |