Assay Design
Gene Transcripts
- Gene Symbol
- OR4F13P
- Entrez Gene ID
-
390651
- Gene Name
- olfactory receptor family 4 subfamily F member 13 pseudogene
- Gene Aliases
-
GPCRLTM7
- Chromosome Location
-
Chr.15: 101842119 - 101850324 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046417.1 |
|
2 - 3 |
112 |
116 |
GenBank mRNA |
AY792621.1 |
- |
2 - 3 |
112 |
116 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor family 4 subfamily F member 15 |
OR4F15 |
|
-23.8kb
|
Sense |
Chr. - 101818187 - 101819125 on Build GRCh38 |
Downstream |
olfactory receptor family 4 subfamily F member 4 |
OR4F4 |
OLA-7501, OR4F18 |
78.6kb
|
Anti-sense |
Chr. - 101922142 - 101923059 on Build GRCh38 |
View Full Product Details