Assay Design



Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
54749
Gene Name
ependymin related 1
Gene Aliases
EPDR, MERP-1, MERP1, UCC1
Chromosome Location
Chr.7: 37920561 - 37951941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242946.1 NP_001229875.2 1 - 2 649 99
GenBank mRNA DQ914439.1 - 1 - 2 270 99

View Full Product Details