Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251901.1 | NP_001238830.1 | 4 - 5 | 498 | 153 |
NM_004233.3 | NP_004224.1 | 4 - 5 | 669 | 153 | |
GenBank mRNA | AK290426.1 | - | 4 - 5 | 531 | 153 |
AK307059.1 | - | 4 - 5 | 338 | 153 | |
BC030830.1 | - | 4 - 5 | 623 | 153 | |
BT019498.1 | - | 4 - 5 | 491 | 153 | |
CR457019.1 | - | 4 - 5 | 491 | 153 | |
Z11697.1 | - | 4 - 5 | 501 | 153 |