Assay Design



Gene Transcripts

Gene Symbol
DDX53
Entrez Gene ID
168400
Gene Name
DEAD-box helicase 53
Gene Aliases
CAGE, CT26
Chromosome Location
Chr.X: 22999961 - 23002089 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182699.3 NP_874358.2 1 - 1 524 84
GenBank mRNA AY039237.1 - 1 - 1 529 84
BC051703.1 - - 509 84
BC067878.1 - 1 - 1 515 84

View Full Product Details