Assay Design
Gene Transcripts
- Gene Symbol
- CYP4F29P
- Entrez Gene ID
-
54055
- Gene Name
- cytochrome P450 family 4 subfamily F member 29, pseudogene
- Gene Aliases
-
4F-se4[6:7:8], C21orf15, CYP4F-se4[6:7:8], CYP4F3LP
- Chromosome Location
-
Chr.21: 13843133 - 13848364 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026755.1 |
|
1 - 2 |
276 |
65 |
GenBank mRNA |
AY040090.1 |
- |
1 - 2 |
276 |
65 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 8069-2 |
MIR8069-2 |
hsa-mir-8069-2 |
-122.5kb
|
Anti-sense |
Chr. - 13724189 - 13724274 on Build GRCh38 |
Downstream |
ankyrin repeat domain 20 family member A11, pseudogene |
ANKRD20A11P |
C21orf81 |
95.4kb
|
Sense |
Chr. - 13943769 - 13980444 on Build GRCh38 |
View Full Product Details