Assay Design
Gene Transcripts
- Gene Symbol
- ZFX-AS1
- Entrez Gene ID
-
100873922
- Gene Name
- ZFX antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 24146225 - 24149654 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046657.1 |
|
1 - 2 |
98 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW237142.1 |
- |
2 - 3 |
|
|
Chr. X - 24146225 - 24149654 [+] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein, X-linked |
ZFX |
ZNF926 |
|
Anti-sense |
Chr. - 24148994 - 24216255 on Build GRCh38 |
Downstream |
SPT20 homolog, SAGA complex component-like 2 |
SUPT20HL2 |
FAM48B1, FAM48B2 |
161.2kb
|
Sense |
Chr. - 24310862 - 24313315 on Build GRCh38 |
View Full Product Details