Assay Design
Gene Transcripts
- Gene Symbol
- AIRN
- Entrez Gene ID
-
100271873
- Gene Name
- antisense of IGF2R non-protein coding RNA
- Gene Aliases
-
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088
- Chromosome Location
-
Chr.6: 160003291 - 160007664 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_047511.1 |
|
1 - 2 |
1181 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MAS1 proto-oncogene, G protein-coupled receptor |
MAS1 |
MAS, MGRA |
-96.9kb
|
Anti-sense |
Chr. - 159906942 - 159908076 on Build GRCh38 |
Downstream |
calcineurin like EF-hand protein 1 pseudogene |
LOC729603 |
|
85.4kb
|
Anti-sense |
Chr. - 160093082 - 160096212 on Build GRCh38 |
View Full Product Details