Assay Design
Gene Transcripts
- Gene Symbol
- CRIP2
- Entrez Gene ID
-
1397
- Gene Name
- cysteine rich protein 2
- Gene Aliases
-
CRIP, CRP2, ESP1
- Chromosome Location
-
Chr.14: 105472938 - 105480170 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073081.1 |
|
2 - 3 |
229 |
110 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
metastasis associated 1 |
MTA1 |
|
-7.5kb
|
Sense |
Chr. - 105419849 - 105470729 on Build GRCh38 |
Downstream |
cysteine rich protein 1 |
CRIP1 |
CRHP, CRIP, CRP-1, CRP1 |
7.9kb
|
Sense |
Chr. - 105486920 - 105488789 on Build GRCh38 |
View Full Product Details