Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270837.1 | NP_001257766.1 | 6 - 7 | 1038 | 118 |
NM_001270841.1 | NP_001257770.1 | 2 - 3 | 231 | 118 | |
NM_001312.3 | NP_001303.1 | 6 - 7 | 594 | 118 | |
NR_073081.1 | 4 - 5 | 395 | 118 | ||
NR_073082.1 | 6 - 7 | 570 | 118 | ||
NR_073083.1 | 5 - 6 | 499 | 118 | ||
NR_073084.1 | 6 - 7 | 1347 | 118 | ||
NR_073085.1 | 6 - 7 | 819 | 118 | ||
GenBank mRNA | AK057837.1 | - | 5 - 6 | 371 | 118 |
AK091845.1 | - | 6 - 7 | 1347 | 118 | |
AK223251.1 | - | 6 - 7 | 506 | 118 | |
AK300092.1 | - | 6 - 7 | 954 | 118 | |
AK311402.1 | - | 6 - 7 | 819 | 118 | |
AK315757.1 | - | 6 - 7 | 546 | 118 | |
BC000434.2 | - | 6 - 7 | 525 | 118 | |
BC001931.1 | - | 6 - 7 | 525 | 118 | |
BC034151.1 | - | 6 - 7 | 575 | 118 | |
BC128101.1 | - | 6 - 7 | 549 | 118 | |
BE271979.1 | - | 6 - 7 | 495 | 119 | |
BQ684851.1 | - | - | 184 | 118 | |
BT019911.1 | - | 6 - 7 | 506 | 118 | |
CN367966.1 | - | 5 - 6 | 457 | 118 | |
D42123.1 | - | 6 - 7 | 532 | 118 | |
HY078632.1 | - | 4 - 5 | 352 | 118 | |
U36190.1 | - | 6 - 7 | 542 | 118 |