Assay Design
Gene Transcripts
- Gene Symbol
- SNORA70E
- Entrez Gene ID
-
100379250
- Gene Name
- small nucleolar RNA, H/ACA box 70E
- Gene Aliases
-
U70E
- Chromosome Location
-
Chr.11: 83041464 - 83041598 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033347.1 |
|
1 - 1 |
46 |
81 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
DNA damage induced apoptosis suppressor |
DDIAS |
C11orf82, noxin |
-106.8kb
|
Anti-sense |
Chr. - 82901695 - 82934659 on Build GRCh38 |
Downstream |
RAB30 antisense RNA 1 (head to head) |
RAB30-AS1 |
|
30.5kb
|
Anti-sense |
Chr. - 83072066 - 83073712 on Build GRCh38 |
View Full Product Details