Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024766.4 | NP_079042.1 | 9 - 10 | 866 | 174 |
XM_011533113.2 | XP_011531415.1 | 7 - 8 | 542 | 174 | |
XM_017004982.1 | XP_016860471.1 | 7 - 8 | 2969 | 174 | |
XM_017004983.1 | XP_016860472.1 | 8 - 9 | 1081 | 174 | |
GenBank mRNA | AK027104.1 | - | 9 - 10 | 806 | 174 |
BC053733.1 | - | 9 - 10 | 793 | 174 |