Assay Design



Gene Transcripts

Gene Symbol
VGLL2
Entrez Gene ID
245806
Gene Name
vestigial like family member 2
Gene Aliases
VGL2, VITO1
Chromosome Location
Chr.6: 117265558 - 117273565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182645.3 NP_872586.1 2 - 3 596 109
XM_005266883.2 XP_005266940.1 2 - 3 413 109
GenBank mRNA AJ578053.1 - 2 - 3 580 109
AK096878.1 - 2 - 3 413 109

View Full Product Details